Assay Details
Gene Symbol: Papd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107333.1 | NP_001100803.1 | 4 - 5 | 470 | 69 | |
XM_006227781.2 | XP_006227843.1 | 4 - 5 | 4220 | 69 | ||
XM_006227782.2 | XP_006227844.1 | 4 - 5 | 857 | 69 | ||
XM_008758690.1 | XP_008756912.1 | 4 - 5 | 587 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
