Assay Details
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017134.3 | NP_058830.2 | 4 - 5 | 616 | 93 | |
GenBank mRNA | BC091158.1 | - | 4 - 5 | 546 | 93 | |
FM120421.1 | - | 2 - 3 | 258 | 93 | ||
FQ209619.1 | - | 1 - 2 | 126 | 93 | ||
FQ218591.1 | - | 4 - 5 | 560 | 93 | ||
FQ219159.1 | - | 4 - 5 | 497 | 93 | ||
J02720.1 | - | 4 - 5 | 497 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
