Assay Details
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017134.3 | NP_058830.2 | 6 - 7 | 817 | 108 | |
GenBank mRNA | BC091158.1 | - | 6 - 7 | 747 | 108 | |
FM120421.1 | - | 4 - 5 | 459 | 108 | ||
FQ209619.1 | - | 3 - 4 | 328 | 108 | ||
FQ219159.1 | - | 6 - 7 | 108 | |||
J02720.1 | - | 6 - 7 | 698 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
