Assay Details
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017134.3 | NP_058830.2 | 7 - 8 | 946 | 148 | |
GenBank mRNA | BC091158.1 | - | 7 - 8 | 876 | 148 | |
FQ209619.1 | - | 4 - 5 | 457 | 148 | ||
FQ219159.1 | - | 7 - 8 | 828 | 149 | ||
J02720.1 | - | 7 - 8 | 827 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
