Assay Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021869.2 | NP_068641.2 | 5 - 6 | 486 | 122 | |
XM_006227691.2 | XP_006227753.1 | 6 - 7 | 1808 | 122 | ||
XM_008758626.1 | XP_008756848.1 | 6 - 7 | 1802 | 122 | ||
GenBank mRNA | AF031430.1 | - | 4 - 5 | 390 | 122 | |
BC085737.1 | - | 5 - 6 | 486 | 122 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758670.1 | XP_008756892.1 | 3 - 4 | 372 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
