Assay Details
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007620.1 | NP_001007621.1 | 4 - 5 | 594 | 94 | |
XM_006251754.1 | XP_006251816.1 | 5 - 6 | 454 | 94 | ||
GenBank mRNA | BC079137.1 | - | 4 - 5 | 594 | 94 | |
FQ212778.1 | - | 4 - 5 | 283 | 94 | ||
FQ214416.1 | - | 4 - 5 | 283 | 94 | ||
FQ234205.1 | - | 4 - 5 | 283 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
