Assay Details
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110810.1 | NP_001104280.1 | 6 - 7 | 1800 | 89 | |
XM_006255355.2 | XP_006255417.1 | 6 - 7 | 1274 | 89 | ||
GenBank mRNA | EU236721.1 | - | 6 - 7 | 1800 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
