Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 3 - 4 | 407 | 84 | |
XM_006255181.2 | XP_006255243.1 | 3 - 4 | 419 | 84 | ||
XM_006255182.2 | XP_006255244.1 | 3 - 4 | 397 | 84 | ||
XM_006255183.2 | XP_006255245.1 | 2 - 3 | 320 | 84 | ||
XM_006255185.2 | XP_006255247.1 | 3 - 4 | 353 | 84 | ||
GenBank mRNA | BC089985.1 | - | 3 - 4 | 407 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
