Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 1 - 2 | 93 | 99 | |
XM_006255181.2 | XP_006255243.1 | 1 - 2 | 105 | 99 | ||
GenBank mRNA | BC089985.1 | - | 1 - 2 | 93 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089985.1, NM_001011926.1, XM_006255181.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
