Assay Details
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013164.1 | NP_037296.1 | 3 - 4 | 336 | 65 | |
GenBank mRNA | AY325179.1 | - | 3 - 4 | 336 | 65 | |
U51583.1 | - | 412 | 65 | |||
U51584.1 | - | 1553 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U51584.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
