Assay Details
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013164.1 | NP_037296.1 | 4 - 5 | 531 | 74 | |
GenBank mRNA | AY325179.1 | - | 4 - 5 | 531 | 74 | |
U51583.1 | - | 607 | 74 | |||
U51584.1 | - | 1748 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U51584.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
