Assay Details
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022934.1 | NP_075223.1 | 5 - 6 | 766 | 114 | |
XM_006238099.1 | XP_006238161.1 | 5 - 6 | 864 | 114 | ||
GenBank mRNA | BC062009.1 | - | 5 - 6 | 723 | 114 | |
FQ216722.1 | - | 5 - 6 | 755 | 114 | ||
U53922.1 | - | 5 - 6 | 766 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
