Assay Details
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012592.2 | NP_036724.1 | 4 - 5 | 465 | 66 | |
GenBank mRNA | BC088401.1 | - | 4 - 5 | 465 | 66 | |
FQ231497.1 | - | 3 - 4 | 307 | 66 | ||
J05031.1 | - | 4 - 5 | 475 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
