Assay Details
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017052.2 | NP_058748.2 | 1 - 2 | 148 | 52 | |
GenBank mRNA | BC088398.1 | - | 1 - 2 | 148 | 52 | |
BC098919.1 | - | 1 - 2 | 95 | 52 | ||
BC128707.1 | - | 1 - 2 | 124 | 52 | ||
FQ209461.1 | - | 1 - 2 | 142 | 52 | ||
FQ210296.1 | - | 1 - 2 | 147 | 52 | ||
FQ211554.1 | - | 1 - 2 | 147 | 52 | ||
FQ234001.1 | - | 1 - 2 | 37 | 52 | ||
X59037.1 | - | 1 - 2 | 88 | 52 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
