Assay Details
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017125.3 | NP_058821.1 | 1 - 2 | 53 | 78 | |
GenBank mRNA | BC063173.1 | - | 1 - 2 | 56 | 78 | |
FQ221333.1 | - | 1 - 2 | 57 | 78 | ||
X61654.1 | - | 1 - 2 | 53 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC063173.1, NM_017125.3, X61654.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
