Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 7 - 8 | 1112 | 83 | |
XM_008770746.1 | XP_008768968.1 | 6 - 7 | 3759 | 83 | ||
GenBank mRNA | BC081810.1 | - | 7 - 8 | 1112 | 83 | |
FQ213323.1 | - | 7 - 8 | 1100 | 83 | ||
M95591.1 | - | 7 - 8 | 1130 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
