Assay Details
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019905.1 | NP_063970.1 | 3 - 4 | 191 | 85 | |
GenBank mRNA | BC059136.1 | - | 3 - 4 | 210 | 85 | |
FQ225322.1 | - | 3 - 4 | 207 | 85 | ||
L13039.1 | - | 2 - 3 | 162 | 85 | ||
S73559.1 | - | 3 - 4 | 193 | 85 | ||
X66871.1 | - | 3 - 4 | 191 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
