Assay Details
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022612.1 | NP_072134.1 | 1 - 2 | 121 | 95 | |
XM_006234985.2 | XP_006235047.2 | 2 - 3 | 502 | 95 | ||
GenBank mRNA | AF136927.1 | - | 1 - 2 | 121 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
