Assay Details
Gene Symbol: Bard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022622.1 | NP_072144.1 | 3 - 4 | 359 | 120 | |
GenBank mRNA | AF182946.1 | - | 3 - 4 | 359 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
