Assay Details
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022705.1 | NP_073196.1 | 5 - 6 | 718 | 149 | |
XM_008771886.1 | XP_008770108.1 | 6 - 7 | 872 | 149 | ||
XM_008771887.1 | XP_008770109.1 | 5 - 6 | 575 | 149 | ||
GenBank mRNA | M16200.1 | - | 5 - 6 | 718 | 149 | |
Y00311.1 | - | 4 - 5 | 453 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
