Assay Details
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022713.2 | NP_073204.2 | 4 - 5 | 463 | 80 | |
XM_006244202.2 | XP_006244264.1 | 4 - 5 | 643 | 80 | ||
GenBank mRNA | BC066665.1 | - | 4 - 5 | 452 | 80 | |
DN934649.1 | - | 4 - 5 | 494 | 80 | ||
M32725.1 | - | 1 - 2 | 104 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
