Assay Details
Gene Symbol: Mst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024352.1 | NP_077328.1 | 6 - 7 | 694 | 99 | |
XM_006243706.1 | XP_006243768.1 | 6 - 7 | 736 | 99 | ||
GenBank mRNA | BC089796.1 | - | 6 - 7 | 698 | 99 | |
X95096.1 | - | 6 - 7 | 694 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
