Assay Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031041.1 | NP_112303.1 | 1 - 2 | 35 | 67 | |
GenBank mRNA | BC085345.1 | - | 1 - 2 | 40 | 67 | |
FQ231337.1 | - | 1 - 2 | 65 | 67 | ||
X65948.1 | - | 1 - 2 | 35 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085345.1, NM_031041.1, X65948.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
