Assay Details
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031560.2 | NP_113748.1 | 5 - 6 | 688 | 67 | |
GenBank mRNA | AF010306.1 | - | 5 - 6 | 657 | 67 | |
BC078793.1 | - | 5 - 6 | 688 | 67 | ||
FQ222438.1 | - | 5 - 6 | 632 | 67 | ||
FQ228691.1 | - | 5 - 6 | 705 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
