Assay Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032066.1 | NP_114455.1 | 10 - 11 | 847 | 115 | |
XM_006234599.2 | XP_006234661.1 | 10 - 11 | 1013 | 114 | ||
GenBank mRNA | BC061543.1 | - | 10 - 11 | 867 | 114 | |
FQ213557.1 | - | 10 - 11 | 882 | 114 | ||
FQ214777.1 | - | 10 - 11 | 911 | 114 | ||
U81186.1 | - | 10 - 11 | 847 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid biosynthetic process
estrogen biosynthetic process
biological_process
positive regulation of cell-substrate adhesion
extracellular matrix organization
fibronectin binding
molecular_function
estradiol 17-beta-dehydrogenase activity
collagen binding
heparin binding
oxidoreductase activity
fatty acid biosynthetic process
estrogen biosynthetic process
biological_process
positive regulation of cell-substrate adhesion
extracellular matrix organization
fibronectin binding
molecular_function
estradiol 17-beta-dehydrogenase activity
collagen binding
heparin binding
oxidoreductase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P517896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13136 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517896 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517896 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P517896 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P517896 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700309975 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97519 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101274 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 425 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48073 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92196 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53959 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60563 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5857 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48113 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4023 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700309975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10847656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847655 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10847646 | - | - |
Rat Genome 230 2.0 Array | 1368051_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368051_at | - | - |
Affymetrix HT Rat Focus Array | 1368051_at | - | - |
Affymetrix HT Rat Focus Array | 1368051_PM_at | - | - |
Rat Expression Set 230 | 1368051_PM_at | - | - |
Rat Expression Set 230 | 1368051_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847646 | - | - |
Rat Genome U34 Set | U81186_at | - | - |
Rat Genome U34 Set | U81186_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372600 | - | - |
RatRef-12 Expression BeadChip | ILMN_51036 | - | - |
