Assay Details
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053582.2 | NP_446034.1 | 6 - 7 | 863 | 66 | |
XM_006238977.2 | XP_006239039.1 | 7 - 8 | 66 | |||
XM_006238978.2 | XP_006239040.1 | 7 - 8 | 66 | |||
GenBank mRNA | AB050717.1 | - | 6 - 7 | 864 | 66 | |
BC097299.1 | - | 7 - 8 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
