Assay Details
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057132.3 | NP_476473.1 | 1 - 2 | 156 | 76 | |
XM_006243699.1 | XP_006243761.1 | 3 - 4 | 424 | 76 | ||
XM_006243700.1 | XP_006243762.1 | 3 - 4 | 399 | 76 | ||
XM_006243701.1 | XP_006243763.1 | 3 - 4 | 519 | 76 | ||
GenBank mRNA | AY026068.1 | - | 1 - 2 | 156 | 76 | |
AY026069.1 | - | 1 - 2 | 156 | 76 | ||
BC061732.1 | - | 3 - 4 | 333 | 76 | ||
D84477.1 | - | 1 - 2 | 126 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
