Assay Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255855.1 | XP_006255917.1 | 2 - 3 | 306 | 90 | |
GenBank mRNA | AF374406.1 | - | 1 - 2 | 138 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
