Assay Details
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134329.1 | NP_599156.1 | 6 - 7 | 888 | 116 | |
XM_008761493.1 | XP_008759715.1 | 7 - 8 | 1275 | 116 | ||
GenBank mRNA | X98746.1 | - | 6 - 7 | 888 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
