Assay Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 13 - 14 | 1069 | 101 | |
XM_006244296.2 | XP_006244358.1 | 13 - 14 | 1127 | 101 | ||
XM_008766751.1 | XP_008764973.1 | 14 - 15 | 1585 | 101 | ||
GenBank mRNA | AF482705.1 | - | 13 - 14 | 1069 | 101 | |
BC086988.1 | - | 13 - 14 | 1080 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
