Assay Details
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 1 - 2 | 92 | 92 | |
XM_008763989.1 | XP_008762211.1 | 2 - 3 | 1394 | 92 | ||
GenBank mRNA | U67139.1 | - | 1 - 2 | 59 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001276304.1, U67139.1, XM_008763989.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
