Assay Details
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182844.1 | NP_878264.1 | 5 - 6 | 648 | 96 | |
XM_006244110.2 | XP_006244172.1 | 5 - 6 | 688 | 96 | ||
XM_008766684.1 | XP_008764906.1 | 6 - 7 | 766 | 96 | ||
XM_008766685.1 | XP_008764907.1 | 7 - 8 | 880 | 96 | ||
XM_008766686.1 | XP_008764908.1 | 5 - 6 | 690 | 96 | ||
XM_008766687.1 | XP_008764909.1 | 4 - 5 | 1161 | 96 | ||
GenBank mRNA | AY331984.1 | - | 5 - 6 | 648 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
