Assay Details
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199103.1 | NP_954534.1 | 7 - 8 | 1032 | 72 | |
XM_006229580.2 | XP_006229642.1 | 9 - 10 | 2280 | 72 | ||
XM_006229584.2 | XP_006229646.1 | 9 - 10 | 1266 | 72 | ||
XM_006229585.2 | XP_006229647.1 | 8 - 9 | 1226 | 72 | ||
XM_008773600.1 | XP_008771822.1 | 8 - 9 | 5340 | 72 | ||
XM_008773601.1 | XP_008771823.1 | 9 - 10 | 2283 | 72 | ||
XM_008773602.1 | XP_008771824.1 | 9 - 10 | 2297 | 72 | ||
XM_008773603.1 | XP_008771825.1 | 9 - 10 | 2288 | 72 | ||
XM_008773604.1 | XP_008771826.1 | 8 - 9 | 5337 | 72 | ||
XM_008773605.1 | XP_008771827.1 | 8 - 9 | 5337 | 72 | ||
XM_008773606.1 | XP_008771828.1 | 8 - 9 | 5334 | 72 | ||
XM_008773607.1 | XP_008771829.1 | 9 - 10 | 5380 | 72 | ||
XM_008773608.1 | XP_008771830.1 | 9 - 10 | 1270 | 72 | ||
XM_008773609.1 | XP_008771831.1 | 9 - 10 | 1478 | 72 | ||
XM_008773610.1 | XP_008771832.1 | 8 - 9 | 1230 | 72 | ||
GenBank mRNA | AY392762.1 | - | 7 - 8 | 1032 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
