Assay Details
Gene Symbol: Clca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201419.1 | NP_958822.1 | 10 - 11 | 1662 | 86 | |
GenBank mRNA | AY333961.1 | - | 10 - 11 | 1662 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
