Assay Details
Gene Symbol: Ephx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034090.3 | NP_001029262.1 | 4 - 5 | 731 | 122 | |
NM_012844.3 | NP_036976.2 | 4 - 5 | 760 | 122 | ||
GenBank mRNA | BC061568.1 | - | 4 - 5 | 691 | 122 | |
M26125.1 | - | 736 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
