Assay Details
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013073.3 | NP_037205.2 | 1 - 2 | 163 | 76 | |
GenBank mRNA | BC088417.1 | - | 1 - 2 | 163 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
