Assay Details
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013161.2 | NP_037293.1 | 5 - 6 | 485 | 76 | |
GenBank mRNA | D88534.1 | - | 4 - 5 | 457 | 76 | |
D88535.1 | - | 4 - 5 | 457 | 76 | ||
M58369.1 | - | 5 - 6 | 486 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
