Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242896.1 | XP_006242958.1 | 1 - 2 | 248 | 73 | |
XM_006242897.1 | XP_006242959.1 | 1 - 2 | 248 | 73 | ||
XM_008766148.1 | XP_008764370.1 | 1 - 2 | 248 | 73 | ||
GenBank mRNA | FQ226062.1 | - | 1 - 2 | 85 | 73 | |
FQ231091.1 | - | 1 - 2 | 146 | 73 | ||
X06827.1 | - | 1 - 2 | 37 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X06827.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
