Assay Details
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013223.1 | NP_037355.1 | 8 - 9 | 802 | 100 | |
GenBank mRNA | BC081838.1 | - | 8 - 9 | 835 | 100 | |
L27707.1 | - | 8 - 9 | 802 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
