Assay Details
Gene Symbol: Cel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016997.2 | NP_058693.2 | 4 - 5 | 561 | 65 | |
GenBank mRNA | M15893.1 | - | 4 - 5 | 561 | 65 | |
X16054.1 | - | 4 - 5 | 557 | 65 | ||
Z22803.1 | - | 3 - 4 | 480 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
