Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 6 - 7 | 2131 | 98 | |
GenBank mRNA | AF284345.1 | - | 6 - 7 | 1828 | 98 | |
AF292389.1 | - | 6 - 7 | 2131 | 98 | ||
DQ137428.1 | - | 1 - 2 | 94 | 98 | ||
U69884.1 | - | 6 - 7 | 1835 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
