Assay Details
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022395.2 | NP_071790.1 | 12 - 13 | 1422 | 78 | |
GenBank mRNA | BC078826.1 | - | 12 - 13 | 1417 | 78 | |
D13907.1 | - | 12 - 13 | 1400 | 78 | ||
L12965.1 | - | 12 - 13 | 1423 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
