Assay Details
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022546.1 | NP_071991.1 | 1 - 2 | 79 | 102 | |
GenBank mRNA | AJ006971.1 | - | 1 - 2 | 79 | 102 | |
BC062076.1 | - | 1 - 2 | 80 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AJ006971.1, BC062076.1, NM_022546.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
