Assay Details
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112712.1 | NP_001106183.1 | 5 - 6 | 451 | 84 | |
NM_001112713.1 | NP_001106184.1 | 5 - 6 | 451 | 84 | ||
NM_031657.3 | NP_113845.2 | 5 - 6 | 451 | 84 | ||
XM_006253653.2 | XP_006253715.1 | 5 - 6 | 560 | 84 | ||
GenBank mRNA | AF040750.1 | - | 5 - 6 | 451 | 84 | |
CV795758.1 | - | 3 - 4 | 84 | |||
EX488847.1 | - | 5 - 6 | 611 | 84 | ||
X97439.1 | - | 5 - 6 | 451 | 84 | ||
X97440.1 | - | 5 - 6 | 451 | 84 | ||
Y09365.1 | - | 5 - 6 | 479 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
