Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 10 - 11 | 1998 | 116 | |
XM_006245576.2 | XP_006245638.1 | 10 - 11 | 2068 | 116 | ||
GenBank mRNA | AF321415.1 | - | 10 - 11 | 1998 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
