Assay Details
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245586.2 | XP_006245648.1 | 2 - 3 | 879 | 150 |
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 2 - 3 | 808 | 150 | |
XM_006245576.2 | XP_006245638.1 | 2 - 3 | 878 | 150 | ||
GenBank mRNA | AF321415.1 | - | 2 - 3 | 808 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
