Assay Details
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139104.1 | NP_620804.1 | 1 - 2 | 123 | 81 | |
XM_006233668.2 | XP_006233730.1 | 1 - 2 | 303 | 81 | ||
GenBank mRNA | AF223678.1 | - | 1 - 2 | 123 | 81 | |
BC078725.1 | - | 1 - 2 | 144 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF223678.1, BC078725.1, NM_139104.1, XM_006233668.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
