Assay Details
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153311.1 | NP_695223.1 | 9 - 10 | 1030 | 91 | |
XM_008766194.1 | XP_008764416.1 | 7 - 8 | 893 | 91 | ||
GenBank mRNA | AF537098.1 | - | 9 - 10 | 1030 | 91 | |
AF537099.1 | - | 7 - 8 | 808 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
