Assay Details
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182824.2 | NP_877976.1 | 1 - 2 | 83 | 132 | |
GenBank mRNA | AY190319.1 | - | 1 - 2 | 86 | 132 | |
BC100064.1 | - | 1 - 2 | 83 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY190319.1, BC100064.1, NM_182824.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
