Assay Details
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040186.2 | NP_001035276.1 | 7 - 8 | 1064 | 89 | |
NM_012811.3 | NP_036943.1 | 6 - 7 | 896 | 89 | ||
GenBank mRNA | BC085754.1 | - | 6 - 7 | 849 | 89 | |
D84068.1 | - | 6 - 7 | 856 | 89 | ||
DQ455823.1 | - | 7 - 8 | 1017 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
